Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.50 µm (28473-U)
oven 40 C (1 min), 12 C/min to 250 C, 25 C/min to 340 C (4.5 min)
carrier gas helium, 1.5 ml/min constant flow
sample 50 ng on-column of a 72 component semivolatile standard and 8 surrogate compounds, plus 6 internal standards (at 40 ng on-column)
injection 1.0 µL, splitless (0.50 min)
liner 4 mm I.D., single taper
inj. temp. 250 C
MSD interface 340 C
scan range m/z = 40-450
Description
Categories Analytical Chromatography, GC Applications, Semivolatiles
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,4-Trichlorobenzene
  • 1,2-Dichlorobenzene
  • 1,2-Dichlorobenzene-d4
  • 1,3-Dichlorobenzene
  • 1,4-Dichlorobenzene
  • 1,4-Dichlorobenzene-d4
  • 2,4,5-Trichlorophenol
  • 2,4,6-Tribromophenol
  • 2,4,6-Trichlorophenol
  • 2,4-Dichlorophenol
  • 2,4-Dimethylphenol
  • 2,4-Dinitrophenol
  • 2,4-Dinitrotoluene
  • 2,6-Dinitrotoluene
  • 2-Chloronaphthalene
  • 2-Chlorophenol
  • 2-Chlorophenol-d4
  • 2-Fluorobiphenyl
  • 2-Fluorophenol
  • 2-Methyl-4,6-dinitrophenol
  • 2-Methylnaphthalene
  • 2-Methylphenol
  • 2-Nitroaniline
  • 2-Nitrophenol
  • 3,3′-Dichlorobenzidine
  • 3,3′-Dimethylbenzidine
  • 3-Nitroaniline
  • 4-Bromophenylphenylether
  • 4-Chloro-3-methylphenol
  • 4-Chloroaniline
  • 4-Chlorophenylphenyl ether
  • 4-Methylphenol
  • 4-Nitroaniline
  • 4-Nitrophenol
  • Acenaphthene
  • Acenaphthene-d10
  • Acenaphthylene
  • Aniline
  • Anthracene
  • Azobenzene
  • Benzidine
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[b]fluoranthene
  • Benzo[g,h,i]perylene
  • Benzo[k]fluoranthene
  • Benzoic acid
  • Benzyl alcohol
  • Bis(2-chloroethoxymethane)
  • Bis(2-chloroethyl) ether
  • Bis(2-chloroisopropyl) ether
  • Bis(2-ethylhexyl) phthalate
  • Butylbenzylphthalate
  • Carbazole
  • Chrysene
  • Chrysene-d12
  • Dibenzo[a,h]anthracene
  • Dibenzofuran
  • Diethyl phthalate
  • Dimethylphthalate
  • Di-n-Butyl phthalate
  • Di-n-Octyl phthalate
  • Fluoranthene
  • Fluorene
  • Hexachlorobenzene
  • Hexachlorobutadiene
  • Hexachlorocyclopentadiene
  • Hexachloroethane
  • Indeno[1,2,3-cd]pyrene
  • Isophorone
  • Naphthalene
  • Naphthalene-d8
  • Nitrobenzene
  • Nitrobenzene-d5
  • N-Nitrosodimethylamine
  • N-Nitroso-di-n-propylamine
  • N-Nitrosodiphenylamine
  • Pentachlorophenol
  • Perylene-d12
  • Phenanthrene
  • Phenanthrene-d12
  • Phenol
  • Phenol-d6
  • Pyrene
  • Pyridine
  • Terphenyl-d12