Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.50 µm (28473-U)
oven 50 C, 25 C/min to 275 C, 5 C/min to 330 C
carrier gas helium, 1.5 ml/min., constant
sample Each analyte at 5 µg/mL in methylene chloride
injection 1.0 µL, pulsed splitless (30 psi until 0.1 min, splitter open at 1 min)
liner 2 mm I.D. straight
inj. temp. 250 C
MSD interface 330 C
scan range m/z 45 - 450
Description
Categories Analytical Chromatography, GC Applications, PAHs, Hydrocarbons, Semivolatiles
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Acenaphthene
  • Acenaphthylene
  • Anthracene
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[b]fluoranthene
  • Benzo[g,h,i]perylene
  • Benzo[k]fluoranthene
  • Chrysene
  • Dibenzo[a,h]anthracene
  • Fluoranthene
  • Fluorene
  • Indeno[1,2,3-cd]pyrene
  • Naphthalene
  • Phenanthrene
  • Pyrene