US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms
US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 C (1 min), 10 C/min to 325 C (5 min)
carrier gas helium, 0.9 ml/min., constant
sample Each analyte at 10 µg/mL in methylene chloride
injection 1 µL, splitless (1 min)
liner 3.8 mm I.D., straight
inj. temp. 300 C
detector FID, 365 C
MSD interface 300 C
scan range m/z 45-450
Description
Categories Analytical Chromatography, GC Applications, PAHs, Hydrocarbons, Semivolatiles
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Acenaphthene
  • Acenaphthylene
  • Anthracene
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[b]fluoranthene
  • Benzo[g,h,i]perylene
  • Benzo[k]fluoranthene
  • Chrysene
  • Dibenzo[a,h]anthracene
  • Fluoranthene
  • Fluorene
  • Indeno[1,2,3-cd]pyrene
  • Naphthalene
  • Phenanthrene
  • Pyrene