US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Conditions
column SLB-5ms, 20 m x 0.18 mm I.D., 0.36 µm (28576-U)
oven 60 °C (0.1 min), 25 °C/min to 300 °C, 5 °C/min to 340 °C
carrier gas helium, 0.8 mL/min, constant
sample each analyte at 5 µg/mL in methylene chloride
injection 1.0 µL, pulsed splitless (30 psi until 0.1 min, splitter open at 1 min)
liner 2 mm I.D. straight
inj. temp. 250 °C
MSD interface 340 °C
scan range m/z 45 - 450
Description
Categories Analytical Chromatography, GC Applications, Semivolatiles, PAHs, Hydrocarbons
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Acenaphthene
  • Acenaphthylene
  • Anthracene
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[b]fluoranthene
  • Benzo[g,h,i]perylene
  • Benzo[k]fluoranthene
  • Chrysene
  • Dibenzo[a,h]anthracene
  • Fluoranthene
  • Fluorene
  • Indeno[1,2,3-cd]pyrene
  • Naphthalene
  • Phenanthrene
  • Pyrene