US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 40 C (2 min), 15 C/min to 240 C, 10 C/min to 330 C (2 min)
carrier gas helium, 1.0 ml/min (11 min), ramp to 1.5 ml/min (constant for remainder of run)
sample 50 ppm of an 80 component semivolatile standard, plus 6 internal standards (at 40 ppm) in methylene chloride
injection 1 µL, pulsed splitless (15 psi until 0.10 min), (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 C
MSD interface 330 C
scan range m/z = 40-450
Description
Categories Analytical Chromatography, GC Applications, Semivolatiles
Featured Industry Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1'-Biphenyl
  • 1,2,4-Trichlorobenzene
  • 1,2-Dichlorobenzene
  • 1,2-Dichlorobenzene-d4
  • 1,3-Dichlorobenzene
  • 1,4-Dichlorobenzene
  • 1,4-Dichlorobenzene-d4
  • 2,4,5-Trichlorophenol
  • 2,4,6-Tribromophenol
  • 2,4,6-Trichlorophenol
  • 2,4-Dichlorophenol
  • 2,4-Dimethylphenol
  • 2,4-Dinitrophenol
  • 2,4-Dinitrotoluene
  • 2,6-Dinitrotoluene
  • 2-Chloronaphthalene
  • 2-Chlorophenol
  • 2-Chlorophenol-d4
  • 2-Fluorobiphenyl
  • 2-Fluorophenol
  • 2-Methyl-4,6-dinitrophenol
  • 2-Methylnaphthalene
  • 2-Methylphenol
  • 2-Nitroaniline
  • 2-Nitrophenol
  • 3,3′-Dichlorobenzidine
  • 3-Nitroaniline
  • 4-Bromophenyl phenyl ether
  • 4-Chloro-3-methylphenol
  • 4-Chlorophenylphenyl ether
  • 4-Methylphenol
  • 4-Nitroaniline
  • 4-Nitrophenol
  • 5-Chloroaniline
  • Acenaphthene
  • Acenaphthene-d10
  • Acenaphthylene
  • Acetophenone
  • Anthracene
  • Atrazine
  • Azobenzene
  • Benzaldehyde
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[b]fluoranthene
  • Benzo[g,h,i]perylene
  • Benzo[k]fluoranthene
  • Benzoic acid
  • Benzyl alcohol
  • Bis(2-chloroethoxy)methane
  • Bis(2-chloroethyl) ether
  • Bis(2-chloroisopropyl) ether
  • Bis(2-ethylhexyl) phthalate
  • Butylbenzylphthalate
  • Caprolactam
  • Carbazole
  • Chrysene
  • Chrysene-d12
  • Dibenzo[a,h]anthracene
  • Dibenzofuran
  • Diethyl phthalate
  • Dimethylphthalate
  • Di-n-Butyl phthalate
  • Di-n-Octyl phthalate
  • Fluoranthene
  • Fluorene
  • Hexachlorobenzene
  • Hexachlorobutadiene
  • Hexachlorocyclopentadiene
  • Hexachloroethane
  • Indeno[1,2,3-cd]pyrene
  • Isophorone
  • Naphthalene
  • Naphthalene-d8
  • Nitrobenzene
  • Nitrobenzene-d5
  • N-Nitroso-di-n-propylamine
  • N-Nitrosodiphenylamine
  • Pentachlorophenol
  • Perylene-d12
  • Phenanthrene
  • Phenanthrene-d10
  • Phenol
  • Phenol-d6
  • Pyrene
  • Terphenyl-d14