US EPA Method TO-17: GC Analysis of Volatiles on VOCOL after Collection/Desorption using Carbotrap® 300 Tube
US EPA Method TO-17: GC Analysis of Volatiles on VOCOL after Collection/Desorption using Carbotrap® 300 Tube
Conditions
sample/matrix JHAP-43 Gas Mix (500429), 1ppm each component, 20mL
adsorbent tube Carbotrap 300 (28283-U)
desorption process 20 mL/min at 330 °C for 6 min (cryo-focused to inlet @ -150C)
column SPB-1, 60 m x 0.25 mm I.D., 3.0 mum
oven 35C (8 min), 5C/min to 100C (0 min), 15C/min to 230C (8 min)
carrier gas helium 31cm/sec
detector MSD, scan range 35-269
Description
Categories Analytical Chromatography, GC Applications, Air Monitoring Applications, Volatiles
Featured Industry Environmental
Industrial Hygiene
General description This is an application, not a product for sale.
See "Related Products" to find associated products for sale.
Or, for more information, email: Supelco Technical Service
Legal Information Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC