NIOSH Method 7903: IC Analysis of Inorganic Acids after Collection/Desorption using ORBO™-53
NIOSH Method 7903: IC Analysis of Inorganic Acids after Collection/Desorption using ORBO™-53
Conditions
sample/matrix 1 ppm of each component
adsorbent tube ORBO-53 (20265)
column Dionex S2 Fast Run Anion, 25 cm x 3 mm I.D.
mobile phase 3 mM sodium bicarbonate; 2.4 mM sodium carbonate
flow rate 3 mL/min
detector conductivity (10muSiemans/ cm)
Description
Categories Analytical Chromatography, Air Monitoring Applications, Inorganic Acids
Featured Industry Industrial Hygiene
General description This is an application, not a product for sale.
See "Related Products" to find associated products for sale.
For more information, email: Supelco Technical Service
Legal Information ORBO is a trademark of Sigma-Aldrich Co. LLC
suitability application for air monitoring, application for HPLC