EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18
EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 5 µm particles (50537-U)
column temp. 42 °C
mobile phase [A] water; [B] methanol; (72:28, A:B)
flow rate 2 mL/min
pressure 3277 psi (226 bar)
sample 0.033 mg/mL in 33/67 water/acetonitrile
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Explosives
Featured Industry Environmental