Fast, High Resolution Size Exclusion using TSKgel® PWXL HPLC Columns
Fast, High Resolution Size Exclusion using TSKgel® PWXL HPLC Columns
Conditions
column TSKgel G4000 PWXL, 30 cm x 7.8 mm I.D., 7 µm particles (808022)
column TSKgel G4000PW, 60 cm x 7.5 mm I.D., 17 µm particles (805107)
column TSKgel G2500 PWXL, 30 cm x 7.8 mm I.D., 7 µm particles (808020)
column TSKgel G2500PW, 60 cm x 7.5 mm I.D., 12 µm particles (808029)
column temp. 50 °C (G2500PWXL and G4000PWXL columns)
column temp. 25 °C (G2500PW and G400PW columns)
mobile phase 0.1 M NaCl (G2500PWXL and G4000PWXL columns)
mobile phase distilled water (G2500PW and G4000PW columns)
flow rate 1 mL/min
sample A and B: polyethylene glycol 200; C and D: polyethylene oxide standards, SE150, SE15, SE2 in 100 µL
detector refractive index
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.