HPLC Analysis of 1,1'-Binaphthyl-2,2'-diyl Hydrogenphosphate Enantiomers on Astec® (R,R) P-CAP™
HPLC Analysis of 1,1'-Binaphthyl-2,2'-diyl Hydrogenphosphate Enantiomers on Astec® (R,R) P-CAP™
Conditions
column Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 µm particles (31024AST)
column temp. 25 °C
mobile phase 20 mM ammonium acetate in: (A) acetonitrile; (B) methanol; (70:30, A:B)
flow rate 0.9 mL/min
sample 2 mg/mL in methanol
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Chemicals, Chemicals and Industrial Polymers, Binaphthyls
Featured Industry Chemicals and Industrial Polymers
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC