HPLC Analysis of 1,5-Napthalenedisulphonic Acid on SeQuant ZIC-HILIC
HPLC Analysis of 1,5-Napthalenedisulphonic Acid on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
column temp. 40 °C
mobile phase [A] acetonitrile; [B] 10 mM ammonium acetate, pH 6.8; (80:20, A:B)
flow rate 0.5 mL/min
injection 10 µL
detector 288 nm
Description
Categories Analytical Chromatography, Organic Acids
Featured Industry Chemicals and Industrial Polymers