HPLC Analysis of 13 Aldehyde/Ketone DNPH Derivatives on Ascentis® Express C18 (High Flow Rate)
HPLC Analysis of 13 Aldehyde/Ketone DNPH Derivatives on Ascentis® Express C18 (High Flow Rate)
Conditions
column Ascentis Express C18, 10 cm x 4.6 mm I.D., 2.7 µm particles (53827-U)
mobile phase [A] water; [B] acetonitrile; (40:60, v/v, A:B)
flow rate 3.5 ml/min
pressure 8296 psi (572 bar)
sample Carb Method 1004 DNPH Mix 2 (47651-U) dilute 3:2 with water
injection 10 µl
detector UV, 360 nm
Description
Categories Analytical Chromatography, Air Monitoring Applications, Aldehydes & Ketones, Aldehydes
Featured Industry Industrial Hygiene
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for air monitoring, application for HPLC