HPLC Analysis of 2-Bromo-3-Methylbutyric Acid Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of 2-Bromo-3-Methylbutyric Acid Enantiomers on Astec® CHIROBIOTIC® R
Conditions
column CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 µm particles (13024AST)
column temp. 25 °C
mobile phase [A] 20 mM ammonium acetate; [B] methanol; (15:85, A:B)(alternate mobile phase of 0.1% ammonium acetate in methanol gives 3.5 and 4.3 min retention)
flow rate 1 mL/min
sample 2 mg/mL in methanol
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Chemicals, Organic Acids, Chemicals and Industrial Polymers, Other Organic Compound Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC