HPLC Analysis of 2-Chloroethanol Content in Hydroxyzine HCl on SeQuant ZIC-HILIC with Refractive Index Detection
HPLC Analysis of 2-Chloroethanol Content in Hydroxyzine HCl on SeQuant ZIC-HILIC with Refractive Index Detection
Conditions
column SeQuant ZIC-HILIC 150 X 4.6 mm, 5.0 uM, 200 Angstrom (1.51455.0001)
column temp. 40 °C
mobile phase [A] 40mM ammonium acetate; [B] acetonitrile;; (25:75, A:B)
flow rate 0.5 mL/min
sample 100 ppm
injection 25 µL
detector Shimadzu Proninance, RI (Detection Range 1, Cell temp. 40 °C
Description
Categories Analytical Chromatography, Pharmaceuticals