Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of 2-Phenylpropionic Acid Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
HPLC Analysis of 2-Phenylpropionic Acid Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
Conditions
column CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 µm particles (24024AST)
column temp. 25 C
mobile phase [A] 10 mM ammonium acetate, pH 4.0; [B] acetonitrile; (75:25, A:B)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 1 µl
detector UV, 235 nm
Description
Categories Analytical Chromatography, Chiral Applications, Chemicals and Industrial Polymers, Organic Acids
Featured Industry Flavors and Fragrances
Food and Beverages
Vitamins, Nutraceuticals, and Natural Products
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC