HPLC Analysis of 3,5-DNB-Tryptophan Methyl Ester Enantiomers on Astec® CYCLOBOND® I 2000 SN
HPLC Analysis of 3,5-DNB-Tryptophan Methyl Ester Enantiomers on Astec® CYCLOBOND® I 2000 SN
Conditions
column CYCLOBOND I 2000 SN, 25 cm x 4.6 mm I.D., 5 µm particles (20524AST)
column temp. 25 °C
mobile phase ethanol
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, 3,5-DNB-Amino Acids, All Amino Acids
Featured Industry Life Science and Biopharma
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC