HPLC Analysis of 3,5-tBu-MeOBIPHEP Enantiomers on Astec® CHIROBIOTIC® V in Normal Phase and Polar Ionic Modes
HPLC Analysis of 3,5-tBu-MeOBIPHEP Enantiomers on Astec® CHIROBIOTIC® V in Normal Phase and Polar Ionic Modes
Conditions
column CHIROBIOTIC® V, 25 cm x 4.6 mm, 5 µm particles (11024AST)
Description
Analysis Note Demonstration of CHIROBIOTIC® column utility in both normal phase and polar ionic modes. The same CHIROBIOTIC® column can be used in all four modes, from aqueous to organic, without memory effects or loss of performance.
Column: CHIROBIOTIC® V, 25 cm x 4.6 mm, 5 μm particles (11024AST). Left: Normal phase mode. 3,5-tBu-MeOBIPHEP enantiomers. n-Hexane:10% ethanol in n-hexane (75:25), 1.5 mL/min. Right: Polar ionic mode. Citalopram enantiomers. Methanol:acetic acid:TEA (99.8:0.1:0.1), 0.5 mL/min.
Categories Analytical Chromatography, Ligands, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC