Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of 3-Phenyllactic Acid Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of 3-Phenyllactic Acid Enantiomers on Astec® CHIROBIOTIC® R
Conditions
column CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 µm particles (13024AST)
column temp. 25 C
mobile phase 0.1% ammonium acetate in methanol
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 0.5 µl
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Chemicals and Industrial Polymers, Organic Acids
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
Vitamins, Nutraceuticals, and Natural Products
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC