HPLC Analysis of 3a,4,5,6-Tetrahydrosuccinimido[3,4-b]acenaphthen-10-one Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of 3a,4,5,6-Tetrahydrosuccinimido[3,4-b]acenaphthen-10-one Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 25 C
mobile phase [A] hexane; [B] ethanol; (75:25, A:B)
flow rate 1 ml/min
sample 4 mg/mL in methanol
injection 1 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Oxygen compounds, Organic Chemicals, Chemicals and Industrial Polymers, Other Organic Compound Applications
Featured Industry Chemicals and Industrial Polymers
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC