HPLC Analysis of 4-Methyl-5-Phenyl-2-Oxazolidone Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of 4-Methyl-5-Phenyl-2-Oxazolidone Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
column temp. 23 °C
mobile phase methanol
flow rate 0.8 mL/min
sample 2 mg/mL in methanol
injection 2 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Bases, Chemicals and Industrial Polymers, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC