HPLC Analysis of 5,5-Dimethyl-4-phenyl-2-oxazolidinone Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of 5,5-Dimethyl-4-phenyl-2-oxazolidinone Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. ambient
mobile phase [A] 0.5% triethylamine acetate, pH 4.1; [B] methanol; (80:20, A:B)
flow rate 1 mL/min
sample 5 mg/mL in methanol
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Bases, Chemicals and Industrial Polymers, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC