HPLC Analysis of 5-Fluorouracil and 5-Fluorouracil Dimer on SeQuant ZIC-HILIC
HPLC Analysis of 5-Fluorouracil and 5-Fluorouracil Dimer on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (80:20, A,B)
flow rate 0.5 mL/min
pressure 418 psi (2.9 MPa)
injection 10 µL
detector 254 nm
Description
Categories Analytical Chromatography, Antineoplastics