Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of 5-Methyl-5-Phenylhydantoin Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of 5-Methyl-5-Phenylhydantoin Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 25 C
mobile phase methanol
flow rate 0.48 ml/min
sample 1 mg/mL in methanol
injection 10 µl
detector UV, 220 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Bases, Chemicals and Industrial Polymers, Amines
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC