HPLC Analysis of 7-Azatryptophan Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of 7-Azatryptophan Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. ambient
mobile phase [A] water; [B] ethanol; (70:30, A:B)
flow rate 0.8 ml/min
sample 5 mg/mL in methanol/water/ammonium hydroxide
injection 5 µl
detector UV, 210 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, Native Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC