HPLC Analysis of a Monoclonal Antibody on Zenix®-C SEC-150 after Partial Proteolysis with IdeS Protease
HPLC Analysis of a Monoclonal Antibody on Zenix®-C SEC-150 after Partial Proteolysis with IdeS Protease
Conditions
column Zenix-C SEC-150, 30 cm X 4.6 mm I.D., 3 µm particles (Z777083)
column temp. ambient
mobile phase 0.15 M potassium phosphate monobasic, pH 7.0 with potassium hydroxide (567329-U)
flow rate 0.35 ml/min
pressure 1450 psi (100 bar)
sample digested SO57 mAb, ~0.5 g/L
injection 1 µl
detector UV, 215 nm
Description
Analysis Note Proteolysis of antibodies can be useful for simplifying further analysis. This application demonstrates separation of the two fragments, F(ab′)2 and scFc from the intact IgG, after partial digestion with IdeS protease.
Categories Analytical Chromatography, Proteins, Peptides
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies