HPLC Analysis of a Monoclonal Antibody on SRT® SEC-300
HPLC Analysis of a Monoclonal Antibody on SRT® SEC-300
Conditions
column SRT SEC-300, 30 cm x 7.8 mm I.D., 5 µm particles (Z777051)
column temp. ambient
mobile phase 150 mM sodium phosphate, pH 7
flow rate 1.0 ml/min
sample MAb 10, 2.5, 0.5 mg/mL
injection 10 µl
Description
Categories Analytical Chromatography, Antibodies
Featured Industry Life Science and Biopharma
Legal Information SRT is a registered trademark of Sepax Technologies