HPLC Analysis of Acebutolol Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Acebutolol Enantiomers on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
column temp. 22 °C
mobile phase [A] methanol; [B] acetic acid; [C] triethylamine; (99.8:0.1:0.1, A:B:C)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 12 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Antihypertensives, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC