Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Acetaminophen, Caffeine, Chlorpheniramine, Ibuprofen, Naproxen, Phenylephrine, and Pseudoephedrine on Ascentis® Express C18
HPLC Analysis of Acetaminophen, Caffeine, Chlorpheniramine, Ibuprofen, Naproxen, Phenylephrine, and Pseudoephedrine on Ascentis® Express C18
Conditions
column Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 µm particles (53822-U)
column temp. 35 C
mobile phase [A] 5 mM ammonium phosphate monobasic, pH 2.0 with phosphoric acid: acetonitrile (98:2); [B] acetonitrile;
gradient 0 to 68% B in 3 min; held at 68% B for 2 min
flow rate 0.4 ml/min
sample 100 µg/mL in 95:5, water: methanol
injection 1 µl
detector UV, 210 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of various over the counter drugs.
Categories Analytical Chromatography, Analgesics