HPLC Analysis of Acrylamide and Acrylic Acid on Discovery® HS F5
HPLC Analysis of Acrylamide and Acrylic Acid on Discovery® HS F5
Conditions
column Discovery HS F5, 15 cm x 4.6 mm I.D., 5 µm particles (567516-U)
column temp. ambient
mobile phase 0.05% formic acid
flow rate 1.0 ml/min
sample 1.0 mg/mL each in mobile phase
injection 1 µl
detector UV, 206 nm
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Organic Acids, Processing/Packaging Contaminants, Acrylamide, Applications (Chromatograms)
Featured Industry Chemicals and Industrial Polymers
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC