HPLC Analysis of Acrylic Acid Derivatives on Discovery® Zr-Carbon
HPLC Analysis of Acrylic Acid Derivatives on Discovery® Zr-Carbon
Conditions
column Discovery Zr-Carbon, 7.5 cm x 2.1 mm I.D., 3 µm particles (65726-U)
column temp. 50 °C
mobile phase [A] 30 mM phosphoric acid, pH 1.9; [B] acetonitrile
flow rate 0.42 mL/min
sample acrylate @ 0.05 g/L, methacrylate @ 0.05 g/L, sulfopropylacrylate @ 0.02 g/L, sulfopropylmethylacrylate @ 0.02 g/L in 30 mM phosphoric acid, pH 1.9
injection 1 µL
detector UV, 210 nm (200 nm will enhance sensitivity ~2x)
Description
Categories Analytical Chromatography, Organic Acids
Featured Industry Chemicals and Industrial Polymers
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC