HPLC Analysis of Acrylic Resin on TSKgel® SuperMultiporeHZ-M vs. TSKgel® SuperHZ
HPLC Analysis of Acrylic Resin on TSKgel® SuperµLtiporeHZ-M vs. TSKgel® SuperHZ
Conditions
column TSKgel(R) SuperµLtiporeHZ-M, 15 cm x 4.6 mm I.D., 4 µm particles (821488)
column temp. 40 °C
mobile phase THF
flow rate 0.35 mL/min
sample acrylic resin, 0.3%
injection 10 µL
detector refractive index (RI)
Description
Categories Analytical Chromatography, Polymers, coatings, and plastics
Featured Industry Chemicals and Industrial Polymers
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.