HPLC Analysis of Active Cannabinoid Compounds in Gummy Bear Candy on Ascentis® Express Biphenyl with UV Detection after Extraction
HPLC Analysis of Active Cannabinoid Compounds in Gummy Bear Candy on Ascentis® Express Biphenyl with UV Detection after Extraction
Conditions
extraction tube SupelQue Non-buffered tube 2 (55295-U)
extraction process Transfer the solution into 50 mL plastic tube (55248-U); add 10 mL acetonitrile; shake for 1 minute by hand; add contents of the Supel QuE Non-Buffered Extraction Tube 2 (55295-U); (shake for 5 minutes using QuEChERS shaker (55278-U); centrifuge for 5 minutes at 5000 rpm; collect top acetonitrile layer; no cleanup is required.)
column Ascentis Express Biphenyl, 10cm x 2.1mm, 2.7 µm particles (64065-U)
column temp. 35 °C
mobile phase (A) 0.1% TFA in water; (B) 0.1% TFA in acetonitrile
gradient 47% B to 50% B in 13 min.; 100% B in 0.1 min; held at 100% B for 3 min; 47% B in 0.1 min; held at 47% B for 2.5 min.
flow rate 0.7 mL/min
pressure 5000 psi (345 bar)
injection 5 µL
detector UV, 220 nm
Description
Analysis Note dissolve in heated water, 1 gummy candy in 20 mL
Categories Analytical Chromatography, Neurotransmitters