HPLC Analysis of Adenosine Monophosphate (AMP) and Adenosine Triphosphate (ATP) on SeQuant ZIC-HILIC
HPLC Analysis of Adenosine Monophosphate (AMP) and Adenosine Triphosphate (ATP) on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 20 x 2.1 mm, 3.5 µm, 100 Ã… (1.50439.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (70:30, A,B)
flow rate 2.0 ml/min
pressure 1886 psi (13.1 MPa)
injection 0.75 µl
detector 254 nm
Description
Categories Analytical Chromatography, Nucleotides