Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Aflatoxin Analogs on Ascentis® C18
HPLC Analysis of Aflatoxin Analogs on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 3 µm particles (581322-U)
column temp. 30 C or 50 C
mobile phase [A] acetonitrile: [B] methanol: [C] water; (20:20:60, A:B:C)
flow rate 1.0 ml/min
sample Aflatoxin Standard Mix, diluted 1:1 with water
injection 20 µl
detector fluorescence, 360 nm excitation, 455 nm emission
Description
Categories Analytical Chromatography, Mycotoxins
Featured Industry Food and Beverages