HPLC Analysis of Aflatoxins on Ascentis® Express C18 with Fluorescence Detection
HPLC Analysis of Aflatoxins on Ascentis® Express C18 with Fluorescence Detection
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 µm particles (53829-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile; (75:25, A:B)
flow rate 2.0 mL/min
pressure 6381 psi (440 bar)
sample 200 µL aflatoxin standard in acetonitrile; 0.5 µg/mL B2 and G2; 2 µg/mL B1 and G1; 800 µL delivatization solution: 70% water, 20% TFA and 10% acetic acidheat for 9 min at 65 °C
injection 50 µL
detector fluorescence, ex 360 nm, em 440 nm
Description
Categories Analytical Chromatography, Mycotoxins
Featured Industry Food and Beverages