HPLC Analysis of Aggregated Antibodies on Zenix® SEC-300, Fast Analysis
HPLC Analysis of Aggregated Antibodies on Zenix® SEC-300, Fast Analysis
Conditions
column Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 µm particles, 300 Ã… (Z777033)
column temp. 25 °C
mobile phase 150 mM sodium phosphate, pH 7
flow rate 1.0 mL/min
sample antibody sample (17 mg/mL)
injection 2.9 µL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Peptides
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies