Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® T (No Mobile Phase Modifier)
HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® T (No Mobile Phase Modifier)
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 C
mobile phase [A] water; [B] acetonitrile; (50:50, A:B)
flow rate 1 ml/min
sample 1 mg/mL in methanol
injection 10 µl
detector UV, 205 nm
Description
Categories Analytical Chromatography, Chiral Applications, Amino Acids, Life Science & Biopharma, All Biochemical Applications, All Amino Acids, Native Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC