HPLC Analysis of Alkaloids on Discovery® RP-AmideC16
HPLC Analysis of Alkaloids on Discovery® RP-AmideC16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 35 C
mobile phase [A] methanol; [B] 25 mM potassium phosphate, dibasic, pH 3.0; (20:80, A:B)
flow rate 2 ml/min
injection 10 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Alkaloids
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC