HPLC Analysis of Allantoin on SeQuant ZIC-HILIC
HPLC Analysis of Allantoin on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
column temp. 30 °C
mobile phase [A] 0.1% (v/v) formic acid; [B] acetonitrile; (25:75, A:B)
flow rate 0.5 mL/min
pressure 232 psi (16 bar)
sample 0.03 mg/ml in Mobile phase
injection 20 µL
detector 210 nm
Description
Categories Analytical Chromatography, Drugs, miscellaneous