HPLC Analysis of Allegra® on Discovery® Cyano
HPLC Analysis of Allegra® on Discovery® Cyano
Conditions
column Discovery Cyano, 15 cm x 4.6 mm I.D., 5 µm particles (59356-U)
column temp. 25 C
mobile phase acetonitrile:25 mM KH2PO4, pH 7.0 (15:85)
flow rate 1 ml/min
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antihistamine, Antihistamines
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC, Allegra is a registered trademark of Chattem