HPLC Analysis of Amides on Discovery® RP-Amide C16
HPLC Analysis of Amides on Discovery® RP-Amide C16
Conditions
column Discovery RP-Amide C16, 15 cm x 4.6 mm, 5 µm particles (505013)
column temp. 30 °C
mobile phase [A] methanol; [B] 25mM monopotassium phosphate, pH 3.0; (20:80, A:B)
flow rate 2 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Analgesics, Amides
Featured Industry Chemicals and Industrial Polymers
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC