Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Amines on Discovery® Zr-PBD at 20° C
HPLC Analysis of Amines on Discovery® Zr-PBD at 20° C
Conditions
column Discovery Zr-PBD, 7.5 cm x 4.6 mm I.D., 3 µm particles (65717-U)
column temp. 20 C
mobile phase [A] 50 mM Triethylammonium Hydroxide (pH 12.6): [B] acetonitrile (75:25, A:B)
flow rate 1 ml/min
injection 1 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Amines
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC