HPLC Analysis of Amines on Discovery® Zr-PS
HPLC Analysis of Amines on Discovery® Zr-PS
Conditions
column Discovery Zr-PS, 5 cm x 4.6 mm I.D., 3 µm particles (65740-U)
column temp. 40 °C
mobile phase [A] 25 mM HCl, pH 1.8; [B]: acetonitrile
flow rate 1 mL/min
sample tripelennamine, triprolidine (1mg/mL), meclizine (3mg/mL)
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Amines
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC