HPLC Analysis of Amino Acids on Ascentis® Si (Silica), HILIC Mode
HPLC Analysis of Amino Acids on Ascentis® Si (Silica), HILIC Mode
Conditions
column Ascentis Si, 15 cm x 2.1 mm I.D., 5 µm particles (581509-U)
column temp. 35 °C
mobile phase [A] 100 mM ammonium formate (pH 3.0, with formic acid), [B] water, [C] acetonitrile
flow rate 0.3 mL/min
sample in 10:90, (50 mM ammonium formate/formic acid, pH 3.0):acetonitrile
injection 2 µL
detector ESI (+), full scan
Description
Categories Analytical Chromatography, Amino Acids