HPLC Analysis of Amino Acids, Polar Neutral and Acidic on Ascentis® Si
HPLC Analysis of Amino Acids, Polar Neutral and Acidic on Ascentis® Si
Conditions
column Ascentis Si, 10 cm x 2.1 mm I.D., 5 µm particles (581508-U)
column temp. 35 C
mobile phase [A] 100 mM ammonium formate, pH 3.0, with formic acid; [B] water; [C] acetonitrile
gradient 5% A constant; 10% B, 85% C to 16% B, 79% C in 20 min
flow rate 0.3 ml/min
sample as indicated, in 10:90, (50 mM ammonium formate/formic acid, (pH 3.0):acetonitrile
injection 2 µl
detector ESI (+), full scan
Description
Categories Analytical Chromatography, Amino Acid Analysis, Application Notes for amino acid analysis, Amino Acids