HPLC Analysis of Amphetamine and Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Amphetamine and Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column Astec CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm (15024AST)
column temp. 40 °C
mobile phase [A] methanol; [B] water; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.1:0.02, A:B:C:D)
flow rate 1 mL/min
pressure 840 psi (58 bar)
sample 100 µg/mL each enantiomer in methanol
injection 5 µL
detector UV, 205 nm
Description
Analysis Note Resolution of the enantiomers of methamphetamine and amphetamine is of clinical interest for two reasons: they exhibit different physiological effects, and it is a means to distinguish between legal and illicit sources of the drugs. Shown here is the rapid and efficient resolution of all four enantiomers on an Astec CHIROBIOTIC V2 column. LC/MS grade solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Categories Analytical Chromatography, Drugs of Abuse
Featured Industry Pharmaceutical (small molecule)
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC