HPLC Analysis of Amphetamines on Discovery® HS F5
HPLC Analysis of Amphetamines on Discovery® HS F5
Conditions
column Discovery HS F5, 5 cm x 2.1 mm I.D., 3 µm particles (567500-U)
column temp. 35 °C
mobile phase [A] 0.1% ammonium acetate in water; [B] 0.1% ammonium acetate in acetonitrile; [C] acetonitrile; (10:30:60, A:B:C)
flow rate 0.2 mL/min
sample 1 ug/mL in acetonitrile
injection 5 µL
detector MS, ESI (+) in Single Ion Recording (SIR) Mode
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Amines
Featured Industry Forensics and Toxicology
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC