HPLC Analysis of Amphetine, Methamphetamine, MDA, and MDMA on Discovery® HS F5
HPLC Analysis of Amphetine, Methamphetamine, MDA, and MDMA on Discovery® HS F5
Conditions
column Discovery HS F5, 15 cm x 4.6 mm I.D., 5 µm particles (567516-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate in [B] water: acetonitrile (10:90)
flow rate 1.5 mL/min
sample 50 µg/mL each (amphetamine, methamphetamine, MDA and MDMA) in methanol
injection 5 µL
detector UV, 210 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, amines, ADHD drugs, Amines, drugs of abuse, AD/HD Drugs
Featured Industry Forensics and Toxicology
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC