HPLC Analysis of α-Amylase (Crude) on TSKgel® Ether-5PW
HPLC Analysis of α-Amylase (Crude) on TSKgel® Ether-5PW
Conditions
column TSKgel Ether-5PW, 7.5 cm x 7.5 mm I.D., 10 µm particles (808641)
mobile phase [A] 0.1 M phosphate buffer + 1.1 M sodium sulfate, pH 7.0 [B] 0.1 M phosphate buffer A to B over 60 min
flow rate 1.0 ml/min
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation