Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of an Energy Drink on Ascentis® Express HILIC, 10 cm (UV Detection)
HPLC Analysis of an Energy Drink on Ascentis® Express HILIC, 10 cm (UV Detection)
Conditions
column Ascentis Express HILIC, 5 cm x 3 mm I.D., 2.7 µm particles (53967-U)
column temp. 35 C
mobile phase [A] 100 mM ammonium acetate, pH 5 (titrated with acetic acid); [B] water; [C] acetonitrile; (9:1:90, A:B:C:)
flow rate 0.6 ml/min
sample Energy Drink Diluted 1:9 in acetonitrile
injection 2 µl
detector UV, 254 nm
Description
Categories Analytical Chromatography, Food & Beverage Analysis, Flavors, Fragrances, Beverage Testing, Sports and Energy Drinks
Featured Industry Food and Beverages